Skip to Content
Merck
All Photos(1)

Key Documents

8.55156

Sigma-Aldrich

NovaSyn TGXV A resin

Novabiochem®

Synonym(s):

NovaSyn TGXV A resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352203
NACRES:
NA.22

Quality Level

product line

NovaSyn® TG
Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-8°C

General description

High-swell version NovaSyn TGA resin (855005) that is designed to have similar properties for Chemmatrix resin. This resin should prove useful for the synthesis of long and aggregated peptides.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service