Skip to Content
Merck
All Photos(1)

Documents

8.55121

Sigma-Aldrich

Wang resin LL (100-200 mesh)

Novabiochem®

Synonym(s):

Wang resin LL (100-200 mesh), p-Benzyloxybenzyl alcohol resin (100-200 mesh) LL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
13111023
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-30°C

General description

A special version of Wang resin with low substitution. Ideal for use in the synthesis of long and difficult peptide acids by Fmoc SPPS.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Linkage

Replaces: 01-64-0471

Analysis Note

Color (visual): white to light yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.30 - 0.50 mmol/g
Swelling Volume (in DMF): lot specific result
Swelling Volume (in CH₂Cl₂): lot specific result
Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g
Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.2 ml/g
The polymer matrix is copoly (styrene-1%DVB), 100 - 200 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service