Skip to Content
Merck
All Photos(1)

Documents

8.55013

Sigma-Aldrich

NovaSyn®TG Sieber resin

Novabiochem®

Synonym(s):

9-Fmoc-amino-xanthen-3-yloxy TG resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

Quality Level

product line

NovaSyn® TG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

NovaSyn TG Sieber resin consists of 130 μm NovaSyn TG bromo resin derivatized with Sieber′s xanhydrylamine linker [1,2]. This resin is ideal for the Fmoc SPPS of partially protected peptide amide fragments. Cleavage from this support is effected by treatment with 1% TFA in DCM, conditions that leave most standard side-chain protecting groups intact. This resin can be reductively alkylated and used to prepare secondary carboxamides [3]. It can also be employed to produce protected peptide fragments in which the C-terminal carboxylic acid group is blocked as a hydroxymethylphenoxy-β-alaninamide ester [4].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] P. Sieber (1987) Tetrahedron Lett., 28, 2107.
[2] C. Somlai, et al. in ′Peptides 1992, Proc.22nd European Peptide Symposium′, C. H. Schneider & A. N. Eberle (Eds), ESCOM, Leiden, 1993, pp. 198.
[3] W. C. Chan, et al. (1995) J. Chem. Soc., Chem. Commun., 1475.
[4] W. C. Chan, et al. (1995) J. Chem. Soc., Chem. Commun., 589.

Linkage

Replaces: 01-64-0092

Analysis Note

Color (visual): light yellow to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.10 - 0.25 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is NovaSyn TG resin (130µm)

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service