8.53008
Boc-His(Dnp)-OH isopropanol
Novabiochem®
Synonym(s):
Boc-His(Dnp)-OH isopropanol, N-α-t.-Boc-N-im-dinitrophenyl-L-histidine isopropanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H19N5O8 · xC3H8O
Molecular Weight:
421.36
UNSPSC Code:
12352209
NACRES:
NA.22
Recommended Products
Quality Level
product line
Novabiochem®
Assay
≥98% (TLC)
form
powder
reaction suitability
reaction type: Boc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
storage temp.
15-25°C
General description
Standard building block for introduction of histidine amino-acid residues by Boc SPPS. The Dnp group is stable to HF and TFMSA. Normally removed with thiophenol prior to HF treatment.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS
Linkage
Replaces: 04-12-0023
Analysis Note
Color (visual): white to yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in EtOAc): +52.5 - +59.5 °
Purity (TLC(011C)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clear soluble
To see the solvent systems used for TLC of Novabiochem® products please click here.
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in EtOAc): +52.5 - +59.5 °
Purity (TLC(011C)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clear soluble
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service