Skip to Content
Merck
All Photos(1)

Key Documents

8.45113

Sigma-Aldrich

Phosphoryl chloride

Msynthplus

Synonym(s):

Phosphoryl chloride, Phosphorus oxide trichloride, Phosphorus oxychloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
153.33
MDL number:
UNSPSC Code:
12352300
EC Index Number:
233-046-7
Assay:
≥99.0% (argenometric)
bp:
105.3 °C/1013 hPa
vapor pressure:
36 hPa ( 20 °C)

vapor pressure

36 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (argenometric)

form

liquid

potency

36 mg/kg LD50, oral (Rat)

pH

1.0 (25 °C, 5 g/L in H2O)

bp

105.3 °C/1013 hPa

mp

1-2 °C

density

1.68 g/cm3 at 20 °C (DIN 51757)

storage temp.

2-30°C

InChI

1S/Cl3OP/c1-5(2,3)4

InChI key

XHXFXVLFKHQFAL-UHFFFAOYSA-N

General description

Msynthplus

Application

Phosphoryl chloride Msynthplus. CAS 10025-87-3, EC Number 233-046-7, chemical formula POCl₃.

Analysis Note

Color (visual)colourlessAppearance of substance (visual)clearAssay (argentometric)≥ 99.0 %Identity (IR)passes testThe analysis of residual solvents is not possible due to the physicochemical properties of the material.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service