Skip to Content
Merck
All Photos(1)

Key Documents

8.41328

Sigma-Aldrich

2-Methylglutaric acid

for synthesis

Synonym(s):

2-Methylglutaric acid, 2-Methylpentanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
MDL number:
UNSPSC Code:
12352106
EC Index Number:
210-521-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.00056 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

form

powder

pH

2.8 (10 g/L in H2O)

mp

74-76 °C

solubility

>1000 g/L

density

1.33 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)C(CCC(=O)O)C

InChI

1S/C6H10O4/c1-4(6(9)10)2-3-5(7)8/h4H,2-3H2,1H3,(H,7,8)(H,9,10)

InChI key

AQYCMVICBNBXNA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 74 °C
Melting range (upper value): ≤ 77 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service