Skip to Content
Merck
All Photos(1)

Key Documents

8.20465

Sigma-Aldrich

Dihexylamine

for synthesis

Synonym(s):

Dihexylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5NH(CH2)5CH3
CAS Number:
Molecular Weight:
185.35
MDL number:
UNSPSC Code:
12352116
EC Index Number:
205-588-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.05 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

250 °C

potency

42 mg/kg LD50, oral (Rat)
<160 mg/kg LD50, skin (Rabbit)

expl. lim.

0.7-5.9 % (v/v)

pH

9.9 (25 °C, 0.3 g/L in H2O)

bp

236 °C/1013 mbar

mp

3 °C

transition temp

flash point 103 °C

density

0.79 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCCCCC)CCCCCC

InChI

1S/C12H27N/c1-3-5-7-9-11-13-12-10-8-6-4-2/h13H,3-12H2,1-2H3

InChI key

PXSXRABJBXYMFT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.786 - 0.787
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service