Skip to Content
Merck
All Photos(1)

Key Documents

8.18839

Sigma-Aldrich

Tetra-n-butylammonium bromide

for synthesis

Synonym(s):

Tetra-n-butylammonium bromide, TBAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4NBr
CAS Number:
Molecular Weight:
322.37
MDL number:
UNSPSC Code:
12352107
EC Index Number:
216-699-2
NACRES:
NA.22

Quality Level

form

crystals

mp

100-103 °C

solubility

600 g/L

density

1.15 g/cm3

bulk density

700 kg/m3

storage temp.

2-30°C

InChI

1S/C16H36N.BrH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

JRMUNVKIHCOMHV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Cobalt-based MOF Nanoribbons: Tetra-n-butylammonium bromide is utilized in the synthesis of cobalt-based metal-organic framework (MOF) nanoribbons, which are applied for the cycloaddition of carbon dioxide to epoxides. This process is crucial for carbon capture and utilization, demonstrating the compound′s role in green chemistry solutions (Ullah et al., 2022).
  • Phosphate Removal: In environmental applications, Tetra-n-butylammonium bromide is incorporated into polyacrylonitrile composite nanofibers to enhance the removal of phosphate from water, showcasing its utility in water treatment technologies (Wang et al., 2021).
  • Iron(III) Spin-crossover Compounds: This quaternary ammonium salt is used in the synthesis of a rare octacoordinated mononuclear iron(III) compound with spin-crossover properties. This application is significant in materials science, particularly in the development of magnetic materials and sensors (Hao et al., 2020).
  • Protein Extraction Technologies: Its application extends to the enrichment and extraction of hydrophobic proteins using fluoroalcohol-induced coacervates. This technique is vital for biochemical research and the pharmaceutical industry, where efficient isolation and purification of proteins are essential (Koolivand et al., 2018).

Analysis Note

Assay (argentometric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service