Skip to Content
Merck
All Photos(1)

Key Documents

8.18613

Sigma-Aldrich

Methyl myristate

for synthesis

Synonym(s):

Methyl myristate, Myristic acid methyl ester, Methyl tetradecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOCH3
CAS Number:
Molecular Weight:
242.40
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-680-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.065 Pa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

bp

295 °C/1001 hPa

mp

18 °C

transition temp

flash point 112 °C

density

0.867 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCCCCCCCCCCCC

InChI

1S/C15H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17-2/h3-14H2,1-2H3

InChI key

ZAZKJZBWRNNLDS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.866 - 0.868
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service