Skip to Content
Merck

8.17076

SAFC

TWEEN® 60 (Polysorbate)

EMPROVE® ESSENTIAL, Ph. Eur., JPE, NF

Pharma Manufacturing

Synonym(s):

Polyoxyethylene sorbitan monostearate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352104
EC Index Number:
500-020-4

Agency

JPE
NF
Ph. Eur.

Quality Level

vapor pressure

<10 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

viscous liquid

technique(s)

API processing | nano-milling: suitable

pH

4,0-7.7 (25 °C, 50 g/L in H2O)

kinematic viscosity

170-320 cSt(40 °C)

transition temp

flash point >149 °C

density

1.1 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharma/biopharma processes
pharmaceutical
semi-solid formulation
solid formulation
solubility enhancement

storage temp.

15-25°C

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Polysorbate 60 is used as surfactant in a variety of liquid formulations and acts e.g. as emulsifying, solubilizing and wetting agent.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service