Skip to Content
Merck
All Photos(1)

Key Documents

8.14927

Sigma-Aldrich

Methyl octanoate

for synthesis

Synonym(s):

Methyl octanoate, Methyl caprylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O2
CAS Number:
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-835-0
NACRES:
NA.22
form:
liquid
Assay:
≥98.0% (GC)

vapor pressure

1.33 hPa ( 34.2 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)

bp

193 °C/1013 hPa

mp

-40 °C

transition temp

flash point 68 °C

density

0.877 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCCCCCC

InChI

1S/C9H18O2/c1-3-4-5-6-7-8-9(10)11-2/h3-8H2,1-2H3

InChI key

JGHZJRVDZXSNKQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.874 - 0.878
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service