Skip to Content
Merck
All Photos(1)

Key Documents

8.14811

Sigma-Aldrich

Tritolyl phosphate

for synthesis

Synonym(s):

Tritolyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21O4P
CAS Number:
Molecular Weight:
368.36
MDL number:
UNSPSC Code:
12352108
EC Index Number:
215-548-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.04 hPa ( 25 °C)

Quality Level

form

liquid

transition temp

flash point >200 °C

density

1.14 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[P](=O)(Oc3ccc(cc3)C)(Oc2ccc(cc2)C)Oc1ccc(cc1)C

InChI

1S/C21H21O4P/c1-16-4-10-19(11-5-16)23-26(22,24-20-12-6-17(2)7-13-20)25-21-14-8-18(3)9-15-21/h4-15H,1-3H3

InChI key

BOSMZFBHAYFUBJ-UHFFFAOYSA-N

Related Categories

Analysis Note

Density (d 20 °C/ 4 °C): 1.165 - 1.185
Identity (IR): passes test

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service