Skip to Content
Merck
All Photos(1)

Key Documents

8.13755

Sigma-Aldrich

Crown ether/Dibenzo-24-crown-8

for synthesis

Synonym(s):

Crown ether/Dibenzo-24-crown-8, Dibenzo-24-crown-8, 1,4,7,10,17,20,23,26-Octaoxa[10.10]orthocyclophane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O8
CAS Number:
Molecular Weight:
448.51
MDL number:
UNSPSC Code:
12352005
EC Index Number:
238-027-7
NACRES:
NA.22

Quality Level

form

powder

mp

100-103 °C

storage temp.

2-30°C

SMILES string

O1CCOCCOc2c(cccc2)OCCOCCOCCOc3c(cccc3)OCC1

InChI

1S/C24H32O8/c1-2-6-22-21(5-1)29-17-13-25-9-10-27-15-19-31-23-7-3-4-8-24(23)32-20-16-28-12-11-26-14-18-30-22/h1-8H,9-20H2

InChI key

UNTITLLXXOKDTB-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99
Identity (IR-spectrum): passes test
Melting range (lower value): 101 - 104
Melting range (upper value): 101 - 105

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service