Skip to Content
Merck
All Photos(1)

Key Documents

8.06912

Sigma-Aldrich

Ethyl mandelate

for synthesis

Synonym(s):

Ethyl mandelate, Ethyl α-hydroxyphenylacetate, Mandelic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COOC2H5
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12352108
EC Index Number:
212-263-0
NACRES:
NA.22
Assay:
≥96.0% (GC)

Quality Level

Assay

≥96.0% (GC)

refractive index

n25/589 1.5136

mp

24-27 °C

transition temp

flash point 93 °C

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)C(O)c1ccccc1

InChI

1S/C10H12O3/c1-2-13-10(12)9(11)8-6-4-3-5-7-8/h3-7,9,11H,2H2,1H3

InChI key

SAXHIDRUJXPDOD-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 25 °C
Melting range (upper value): ≤ 30 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service