Skip to Content
Merck
All Photos(1)

Key Documents

8.04772

Sigma-Aldrich

Iodine trichloride

for synthesis

Synonym(s):

Iodine trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl3I
CAS Number:
Molecular Weight:
233.26
MDL number:
UNSPSC Code:
12352307
EC Index Number:
212-739-8
NACRES:
NA.22

Pricing and availability is not currently available.

vapor pressure

1.3 hPa ( 64 °C)

Quality Level

form

crystals

mp

>60 °C (decomposition)

density

3.12 g/cm3 at 20 °C

SMILES string

[I](Cl)(Cl)Cl

InChI

1S/Cl3I/c1-4(2)3

InChI key

PAWIVBWALDNUJP-UHFFFAOYSA-N

Application

  • Ullmann Reactions of Carbon Nanotubes: This study discusses the use of Ullmann-type reactions for the functionalization of carbon nanotubes, employing iodine trichloride as a chlorinating agent to achieve tunable surface chemistry (A Kolanowska, et al., 2019).
  • Sidewall chlorination of carbon nanotubes by iodine trichloride: This research explores the mechanism of sidewall chlorination of carbon nanotubes using iodine trichloride, highlighting its effectiveness and potential applications in materials science (VK Abdelkader, et al., 2014).
  • An Energy-Dense and High-Power Li-Cl2 Battery by Reversible Interhalogen Bonds: The study introduces iodine trichloride as a cathode material for a Li-Cl2 battery, discussing its role in facilitating oxidation via a liquid-solid pathway and enhancing battery performance (P Li, et al., 2022).
  • Halogen bonding in organic synthesis and organocatalysis: This review covers the application of iodine trichloride as a catalyst for the ring-opening polymerization in organic synthesis (D Bulfield, SM Huber, 2016).

Analysis Note

Assay (iodometric): ≥ 97.0 %

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service