Skip to Content
Merck
All Photos(1)

Key Documents

71205-M

Millipore

Benzonase® Nuclease HC

Purity > 90%, Serratia marcescens, buffered aqueous glycerol solution

Synonym(s):

Endonuclease from Serratia marcescens

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Pricing and availability is not currently available.

Product Name

Benzonase® Nuclease HC, Purity > 90%,

biological source

Serratia marcescens

Quality Level

recombinant

expressed in E. coli

Assay

>90% (SDS-PAGE)

form

buffered aqueous glycerol solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze

concentration

≥250 units/μL

application(s)

research use

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Benzonase® Nuclease is a genetically engineered endonuclease from Serratia marcescens. It degrades all forms of DNA and RNA (single stranded, double stranded, linear and circular) while having no proteolytic activity. It is effective over a wide range of conditions and possesses an exceptionally high specific activity. The enzyme completely digests nucleic acids to 5′-monophosphate terminated oligonucleotides 2 to 5 bases in length (below the hybridization limit), which is ideal for removal of nucleic acids from recombinant proteins, enabling compliance with FDA guidelines for nucleic acid contamination. The ability of Benzonase to rapidly hydrolyze nucleic acids makes the enzyme an excellent choice for viscosity reduction to reduce processing time and increase yields of protein. For example, the enzyme is compatible with BugBuster® and PopCulture® Protein Extraction Reagents and can therefore be added along with these reagents to eliminate viscosity and remove nucleic acids from E. coli extracts.

The enzyme consists of two subunits of 30 kDa each. It is functional between pH 6 and 10 and from 0-42 °C and requires 1-2 mM Mg2+ for activation. The enzyme is also active in the presence of ionic and non-ionic detergents, reducing agents, PMSF (1 mM), EDTA (1 mM) and urea (relative activity depends on specific conditions). Activity is inhibited by > 150 mM monovalent cations, > 100 mM phosphate, > 100 mM ammonium sulfate, or > 100 mM guanidine HCl.

Benzonase Nuclease is available in ultrapure (> 99% by SDS-PAGE) and pure (> 90%) grades at a standard concentration of 25-29 U/μl and at a high concentration (HC) of 250 U/μl. Both preparations are free of detectable protease and have specific activity > 1 × 106 U/mg protein. The > 99% purity grade is tested for endotoxins and contains < 0.25 EU/1000 units. The product is supplied as a 0.2 μm filtered solution in 50% glycerol. Store at -20°C.

Total endotoxin : < 0.25 EU/1,000 units. Purity : > 90% by SDS-PAGE.

Application

Used for the removal of nucleic acid from protein samples.

Biochem/physiol Actions

Digests native or heat-denatured DNA and RNA.

Warning

Toxicity: Standard Handling

Unit Definition

One unit is defined as the amount of enzyme that causes a A260 of 1.0 in 30 minutes, which corresponds to complete digestion of 37 μg DNA.

Legal Information

BUGBUSTER is a registered trademark of Merck KGaA, Darmstadt, Germany
Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
POPCULTURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service