Skip to Content
Merck
All Photos(2)

Key Documents

662142

Sigma-Aldrich

DUB Inhibitor VI, P22077

The DUB Inhibitor VI, P22077 controls the biological activity of DUB. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

DUB Inhibitor VI, P22077, P022077, 1-(5-(2,4-Difluorophenylthio)-4-nitrothiophen-2-yl)ethanone, USP7 Inhibitor II, USP47 Inhibitor II, USP7 Inhibitor II, USP47 Inhibitor II, P022077, 1-(5-(2,4-Difluorophenylthio)-4-nitrothiophen-2-yl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7F2NO3S2
CAS Number:
Molecular Weight:
315.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C1=CC([N+]([O-])=O)=C(SC2=CC=C(F)C=C2F)S1)=O

InChI key

RMAMGGNACJHXHO-UHFFFAOYSA-N

General description

A cell-permeable thiophenylethanone that is shown to inhibit USP7 and USP47 deubiquitinase activity (IC50 = 8.01 & 8.74 µM, respectively), while exhibiting much reduced affinity toward ATXN3, BAP1, and USP1, and displaying no inhibitory activity toward other known PR-619 (Cat. No. 662141) targets (IC50 >50 µM). Both P22077 and PR-619 are shown to increase overall protein polyubiquitination in HEK293T cells in a dose- and time-dependent manner (20 to 150 µM; 0.5 to 20 h), however PR619 treatment results in upregulation of both K48- and K63-linked, while P22077 exposure results in mainly enrichment of K48-linked polyUb chains.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Altun, M., et al. 2011. Chem. Biol.18, 1401.
Tian, X., et al. 2011. Assay Drug Dev. Technol.9, 165.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Franz et al.
Cell reports, 37(2), 109819-109819 (2021-10-14)
The AAA+ ATPase VCP regulates the extraction of SUMO and ubiquitin-modified DNA replication factors from chromatin. We have previously described that active DNA synthesis is associated with a SUMO-high/ubiquitin-low environment governed by the deubiquitylase USP7. Here, we unveil a functional
Sandhini Saha et al.
Cell death & disease, 13(6), 563-563 (2022-06-23)
Nutrient surplus and consequent free fatty acid accumulation in the liver cause hepatosteatosis. The exposure of free fatty acids to cultured hepatocyte and hepatocellular carcinoma cell lines induces cellular stress, organelle adaptation, and subsequent cell death. Despite many studies, the
Joonyoung Her et al.
The Journal of clinical investigation, 134(10) (2024-03-26)
The mammalian SUMO-targeted E3 ubiquitin ligase Rnf4 has been reported to act as a regulator of DNA repair, but the importance of RNF4 as a tumor suppressor has not been tested. Using a conditional-knockout mouse model, we deleted Rnf4 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service