Skip to Content
Merck
All Photos(1)

Key Documents

565740

Sigma-Aldrich

α-Amyloid Precursor Protein Modulator

≥95% (HPLC), solid, PKC activator, Calbiochem®

Synonym(s):

α-Amyloid Precursor Protein Modulator, (2S,5S)-(E,E)-8-(5-(4-(Trifluoromethyl)phenyl)-2,4-pentadienoylamino)benzolactam, PKC Activator V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30F3N3O3
CAS Number:
Molecular Weight:
501.54
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

α-Amyloid Precursor Protein Modulator, A cell-permeable benzolactam derived PKC activator (Ki = 11.9 nM for PKCα) that efficiently enhances non-amyloidogenic α-processing of amyloid precursor protein (APP) even at 100 nM in human fibroblast AG06848.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

methanol: 1 mg/mL
DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

FC(F)(F)c1ccc(cc1)\C=C\C=C\C(=O)Nc2cc3c(cc2)N([C@H](C(=O)N[C@@H](C3)CO)C(C)C)C

InChI

1S/C27H30F3N3O3/c1-17(2)25-26(36)32-22(16-34)15-19-14-21(12-13-23(19)33(25)3)31-24(35)7-5-4-6-18-8-10-20(11-9-18)27(28,29)30/h4-14,17,22,25,34H,15-16H2,1-3H3,(H,31,35)(H,32,36)/b6-4+,7-5+/t22-,25-/m0/s1

InChI key

WOLVEMPZUIFSII-IHHOKICGSA-N

General description

A cell-permeable PKC activator (Ki = 11.9 nM for PKCα) that efficiently enhances non-amyloidogenic α-processing of amyloid precursor protein (APP) but lacks any tumor promoting activity.
A cell-permeable benzolactam derived PKC activator (Ki = 11.9 nM for PKCα) that efficiently enhances non-amyloidogenic α-processing of amyloid precursor protein (APP) even at 100 nM in human fibroblast AG06848. Displays a marked loss of tumor promoting activity.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product does not compete with ATP.
Reversible: no
Target Ki: 11.9 nM for activating PKCα

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kozikowski, A.P., et al. 2003. J. Med. Chem.46, 364.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Jiménez et al.
Cell reports methods, 3(7), 100512-100512 (2023-08-03)
Time-specific modulation of gene expression during differentiation by transcription factors promotes cell diversity. However, estimating their dynamic regulatory activity at the single-cell level and in a high-throughput manner remains challenging. We present FateCompass, an integrative approach that utilizes single-cell transcriptomics
Luiza Ghila et al.
Methods in molecular biology (Clifton, N.J.), 2454, 327-349 (2021-04-01)
Pancreatic islet endocrine cells generated from patient-derived induced pluripotent stem cells represent a great strategy for both disease modeling and regenerative medicine. Nevertheless, these cells inherently miss the effects of the intricate network of systemic signals characterizing the living organisms.
Yan Fung Wong et al.
Nature cell biology (2023-01-24)
Cell proliferation is fundamental for almost all stages of development and differentiation that require an increase in cell number. Although cell cycle phase has been associated with differentiation, the actual process of proliferation has not been considered as having a
Protocol development to further differentiate and transition stem cell-derived pancreatic progenitors from a monolayer into endocrine cells in suspension culture.
Braam, et al.
Scientific Reports, 13, 8877-8877 (2023)
Nina Sofi Funa et al.
Stem cell reports, 19(7), 973-992 (2024-06-29)
Genetic differences between pluripotent stem cell lines cause variable activity of extracellular signaling pathways, limiting reproducibility of directed differentiation protocols. Here we used human embryonic stem cells (hESCs) to interrogate how exogenous factors modulate endogenous signaling events during specification of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service