Skip to Content
Merck
All Photos(1)

Key Documents

557324

Sigma-Aldrich

Riluzole

A cell-permeable benzothiazole compound that potently inhibits glutamate release and blocks Na+ channels.

Synonym(s):

Riluzole, 2-Amino-6-(trifluoromethoxy)benzothiazole, PK 26124

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5F3N2OS
CAS Number:
Molecular Weight:
234.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 8223

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

HCl: 0.1 N
DMSO: 40 mg/mL

cation traces

heavy metals: ≤20 ppm

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)Oc1cc2[s]c(nc2cc1)N

InChI

1S/C8H5F3N2OS/c9-8(10,11)14-4-1-2-5-6(3-4)15-7(12)13-5/h1-3H,(H2,12,13)

InChI key

FTALBRSUTCGOEG-UHFFFAOYSA-N

General description

A cell-permeable benzothiazole compound that potently inhibits glutamate release and blocks Na+ channels. Reported to function as a polyglutamine aggregation inhibitor and stimulate the synthesis of nerve growth factor (NGF), brain-derived neurotrophic factor (BDNF), and glial cell-derived neurotrophic factor (GDNF) in mouse astrocyte cultures. Offers neuroprotection, slows disease progression in amyotrophic lateral sclerosis (ALS) patients, and prolongs survival in animal models of Huntington′s disease (HD).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Inhibits glutamate release and blocks Na+ channels
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Heiser, V., et al. 2002. Proc. Natl. Acad. Sci. USA99, 16400.
Schiefer, J., et al. 2002. Mov. Disord.17, 748.
Mizuta, I., et al. 2001. Neurosci. Lett.310, 117.
Louvel, E., et al. 1997. Trends Pharmacol. Sci.18, 196.
Mizoule, J., et al. 1985. Neuropharmacology24, 767.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken-Ichi Dewa et al.
Nature communications, 15(1), 458-458 (2024-02-02)
In the central nervous system, astrocytes enable appropriate synapse function through glutamate clearance from the synaptic cleft; however, it remains unclear how astrocytic glutamate transporters function at peri-synaptic contact. Here, we report that Down syndrome cell adhesion molecule (DSCAM) in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service