Skip to Content
Merck
All Photos(1)

Key Documents

540213

Sigma-Aldrich

PTP Inhibitor V, PHPS1

The PTP Inhibitor V, PHPS1, also referenced under CAS 314291-83-3, controls the biological activity of PTP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PTP Inhibitor V, PHPS1, 4-(Nʹ-(3-(4-Nitrophenyl)-5-oxo-1-phenyl-1,5-dihydro-pyrazol-(4Z)-ylidene)-hydrazino)-benzenesulfonic acid, PhenylHydrazonoPyrazolone Sulfonate 1, PTPNN1 Inhibitor, EC-PTP/PCPTP1 Inhibitor I, MptpA Inhibitor, PTP-SL, PhenylHydrazonoPyrazolone Sulfonate 1, PTPNN1 Inhibitor, 4-(Nʹ-(3-(4-Nitrophenyl)-5-oxo-1-phenyl-1,5-dihydro-pyrazol-(4Z)-ylidene)-hydrazino)-benzenesulfonic acid, EC-PTP/PCPTP1 Inhibitor I, MptpA Inhibitor, PTP-SL Inhibitor I, PTP1B Inhibitor V, SHP1 Inhibitor VI, SHP2 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15N5O6S
CAS Number:
Molecular Weight:
465.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

reddish

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable phosphotyrosine mimetic that acts as a reversible, active-site targeting, substrate-competitive inhibitor of Shp-2 (IC50 and Ki = 2.1 and 0.73 µM, respectively). PHPS1 inhibits ECPTP, PTP1B, Shp1, mycobacterium MptpA only at higher concentrations (IC50 = 5.4, 19, 30, and 39 µM, respectively) and exhibits little activity against PTPH1, STEP, PTPN7, PTPRK, GLEPP1, or LAR2 even at concentrations as high as 50 µM. Shown to inhibit Shp-2-dependent cellular signaling and tumor cell colonies formation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Hellmuth, K., et al. 2008. Proc. Natl. Acad. Sci. USA105, 7275.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service