Skip to Content
Merck
All Photos(1)

Key Documents

535142

Millipore

Protease Inhibitor Cocktail I

Animal-Free, lyophilized, for the inhibition of serine, esterases, cysteine, trypsin-like and metalloproteases. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Animal-Free Protease Inhibitor Mix, Protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
form:
lyophilized
storage temp.:
−20°C

Product Name

Protease Inhibitor Cocktail Set I, Animal-Free, The Protease Inhibitor Cocktail Set I, Animal-Free controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

shipped in

wet ice

storage temp.

−20°C

General description

A cocktail of five protease inhibitors for the inhibition of a broad range of proteases and esterases. Each vial, when reconstituted with 1 ml H2O, yields a 100X stock solution. When diluted to 1X the cocktail contains 500 µM AEBSF, HCl (Cat. No. 101500), 150 nM Aprotinin (Cat. No. 616371), 1 µM E-64 (Cat. No. 324890), 0.5 mM EDTA, Disodium Salt and 1 µM Leupeptin Hemisulfate (Cat. No. 108975). Available as 1 ml or as a set of 10 x 1 ml.

Application

Protease Inhibitor Cocktail Set I, Animal-Free has been used:
  • as a supplement in cell lysis buffer to extract proteins from non-small-cell lung carcinoma (NSCLC) cell lines
  • in NP-40 buffer to lyse the cells for sodium dodecyl sulfate-12% polyacrylamide gel electrophoresis (SDS-PAGE) and western blotting
  • as a component in lysis buffer to lyse transfected Huh7 cells and to wash the pellets to detect hepatitis B virus (HBV) C protein phosphorylation in the cell

Biochem/physiol Actions

Protease inhibitor cocktails and specific protease inhibitors are valuable resources for scientists involved in proteomic studies, such as protein expression, characterization, biomarker discovery, post-translational modification mapping, protein profiling, and quantitative protein measurement. By adding a protease inhibitor cocktail or specific inhibitor, the integrity of proteins is preserved for subsequent analysis and further sample characterization.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Reconstitute each vial in 1 ml H₂O to yield a 100X stock solution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ca2+/Calmodulin-dependent protein kinase II inhibits hepatitis B virus replication from cccDNA via AMPK activation and AKT/mTOR suppression
Kim J, et al.
Microorganisms, 10(3), 498-498 (2022)
Detection of HBV C Protein Phosphorylation in the Cell
Jung J and Kim K
Bio-protocol, 5(15), e1551-e1551 (2015)
Rongxuan Zhu et al.
Frontiers in oncology, 12, 848483-848483 (2022-04-26)
Rapid tumor growth inevitably results in energy stress, including deficiency of glutamine, a critical amino acid for tumor cell proliferation. However, whether glutamine deficiency allows tumor cells to use lipid droplets as an energy resource and the mechanism underlying this
Waqas Azeem et al.
Frontiers in immunology, 11, 438-438 (2020-04-16)
The transcription factor β-catenin is able to induce tolerogenic/anti-inflammatory features in different types of dendritic cells (DCs). Monocyte-derived dendritic cells (moDCs) have been widely used in dendritic cell-based cancer therapy, but so far with limited clinical efficacy. We wanted to
Yumeng Yang et al.
International journal of molecular sciences, 25(15) (2024-08-10)
Enhancing stalk strength is a crucial strategy to reduce lodging. We identified a maize inbred line, QY1, with superior stalk mechanical strength. Comprehensive analyses of the microstructure, cell wall composition, and transcriptome of QY1 were performed to elucidate the underlying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service