Skip to Content
Merck
All Photos(1)

Key Documents

529531

Sigma-Aldrich

PICK1 PDZ Domain Inhibitor, FSC231

The PICK1 PDZ Domain Inhibitor, FSC231 controls the biological activity of PICK1. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

PICK1 PDZ Domain Inhibitor, FSC231, ( E)-Ethyl-2-cyano-3-(3,4-dichlorophenyl)acryloylcarbamate, (E)-Ethyl-2-cyano-3-(3,4-dichlorophenyl)acryloylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10Cl2N2O3
Molecular Weight:
313.14
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable acryloylcarbamate compound that selectively targets the PDZ (PSD-95/Discs-large/ZO-1 homology) domain of PICK1 (protein interacting with C kinase 1), but not PDZ domains of PSD-95 (postsynaptic density protein 95) and GRIP1 (glutamate receptor interacting protein 1), effectively competing against dopamine transporter/DAT (Ki ~10 µM in competitive binding assays), GluR2 (Ki ~10 µM in competitive binding assays), and mRluR7a (~70% inhibition by Co-IP using lysates from 50 µM FSC231-treated HEK293), c-terminus binding to PICK1 PDZ. Shown to accelerate GluR2 surface recycling after NMDR-induced internalization (t1/2 = 7.5 vs 10 min with and without 50 µM FSC231 treatment) in rat hippocampal neurons and prevent both LTD (% long-term depression = 74 vs 50 with and without 50 µM FSC231 treatment) and LTP (% long-term potentiation = 137 vs 266 with and without 50 µM FSC231 treatment) in murine hippocampal slices.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconsitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Thorsen, T.S., et al. 2010. Proc. Natl. Acad. Sci. USA107, 413.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Dou et al.
BioMed research international, 2021, 9884297-9884297 (2021-07-27)
We performed in vitro and in vivo experiments to explore the role of protein kinase C-binding protein 1 (PICK1), an intracellular transporter involved in oxidative stress-related neuronal diseases, in sepsis-related acute kidney injury (AKI). Firstly, PCR, western blotting, and immunohistochemistry
Zengyan Zhu et al.
Psychopharmacology, 240(1), 239-248 (2022-12-24)
Muscarinic acetylcholine receptors (mAChRs) have been shown to play significant roles in the regulation of normal cognitive processes in the hippocampus, and α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptors (AMPARs) are also involved in these processes. This study aims to explore the mAChR-mediated regulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service