Skip to Content
Merck
All Photos(1)

Key Documents

506154

Sigma-Aldrich

Pifithrin-α, p-Nitro, Cyclic

A cell-permeable p53 inhibitor that exhibits 10-fold higher potency and 50% longer half-life than Pifithrin-α.

Synonym(s):

Pifithrin-α, p-Nitro, Cyclic, 2-(4-Nitrophenyl)imidazo[2,1-b]-5,6,7,8-tetrahydrobenzothiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O2S
CAS Number:
Molecular Weight:
299.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 2.5 mg/mL
ethanol: 250 μg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[s]1c2[n](c4c1CCCC4)cc(n2)c3ccc(cc3)[N+](=O)[O-]

InChI

1S/C15H13N3O2S/c19-18(20)11-7-5-10(6-8-11)12-9-17-13-3-1-2-4-14(13)21-15(17)16-12/h5-9H,1-4H2

InChI key

XMFNSEDROOHGBY-UHFFFAOYSA-N

General description

A cell-permeable p53 inhibitor that exhibits 10-fold higher potency (ED50 = 30 nM in protecting etoposide-induced cortical neuron death) and 50% longer half-life (t1/2 = 6h in neuron culture medium at 37°C) than Pifithrin-α (Cat. No. 506132). However, despite its in vitro efficacy, this inhibitor is not effective when adminstered in rats in vivo. For in vivo applications, please consider Pifithrin-α, p-Nitro (Cat. No. 506152).

Biochem/physiol Actions

Cell permeable: yes
ED50 = 30 nM in protecting etoposide-induced cortical neuron death
Primary Target
p53
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pietrancosta, N., et al. 2006. J. Med. Chem.49, 3645.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard A Zager et al.
American journal of physiology. Renal physiology, 316(4), F674-F681 (2019-01-31)
The cyclin kinase inhibitor p21 is acutely upregulated during acute kidney injury (AKI) and exerts cytoprotective effects. A proposed mechanism is oxidant stress-induced activation of p53, the dominant p21 transcription factor. Glycerol-induced rhabdomyolysis induces profound renal oxidant stress. Hence, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service