Skip to Content
Merck
All Photos(1)

Key Documents

5.05156

Millipore

SUMOylation Inhibitor III, 2-D08

Synonym(s):

SUMOylation Inhibitor III, 2-D08, 2-(2,3,4-Trihydroxyphenyl)-4H-chromen-4-one, 2D08, SUMO E2 Ubc9 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
Molecular Weight:
270.24
UNSPSC Code:
41100000
NACRES:
NA.24

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-brown

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

General description

A cell-permeable trihydroxyflavone compound that inhibits androgen receptor peptide, IκBα, and Topoisomerase I SUMOylation in cell-free assays (>90% at 30 µM) by selectively preventing the transfer of SUMO from the E2 thioester to the substrate without affecting SUMO-activating enzyme E1 or E2 Ubc9-SUMO thioester formation. A 6 h pretreatment with 100 µM 2-D08 is shown to effectively inhibit 10 µM CPT- (Camptothecin; Cat. No. 208925) induced cellular Topoisomerase I SUMOylation in breast cancer ZR-75-1 and BT-474 cultures without affecting overall cellular protein ubiquitinations. Anacardic and Ginkgolic Acid (Cat. No. 172050 and 345887, respectively) in comparison, both inhibit E1- SUMO thioester formation instead.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
SUMO E2-thioester complex
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Kim, Y.S., et al. 2013. Chem. Biol.20, 604.
Cotelle, N., et al. 1996. Free Radic. Biol. Med.20, 35.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service