Skip to Content
Merck
All Photos(1)

Documents

5.04712

Sigma-Aldrich

β-Catenin/Tcf Inhibitor VI, ICG-001

Synonym(s):

β-Catenin/Tcf Inhibitor VI, ICG-001, (6 S,9a S)-Hexahydro-6-((4-hydroxyphenyl)methyl)-8-(1-naphthalenylmethyl)-4,7-dioxo-N-(phenylmethyl)-2H-pyrazino[1,2-a]pyrimidine-1(6H)-carboxamide, (6 S)-N-Benzyl-6-(p-hydroxyphenylmethyl)-8-(1-naphthylmethyl)-4,7-dioxohexahydro-2H-, (6S,9aS)-Hexahydro-6-((4-hydroxyphenyl)methyl)-8-(1-naphthalenylmethyl)-4,7-dioxo-N-(phenylmethyl)-2H-pyrazino[1,2-a]pyrimidine-1(6H)-carboxamide, (6S)-N-Benzyl-6-(p-hydroxyphenylmethyl)-8-(1-naphthylmethyl)-4,7-dioxohexahydro-2H-pyra

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H32N4O4
CAS Number:
Molecular Weight:
548.63
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C33H32N4O4/c38-27-15-13-23(14-16-27)19-29-32(40)35(21-26-11-6-10-25-9-4-5-12-28(25)26)22-30-36(18-17-31(39)37(29)30)33(41)34-20-24-7-2-1-3-8-24/h1-16,29-30,38H,17-22H2,(H,34,41)/t29-,30+/m0/s1

InChI key

HQWTUOLCGKIECB-XZWHSSHBSA-N

General description

A cell-permeable pyrazinopyrimidinecarboxamide that competes against β-catenin for CBP (CREB-binding protein) binding (5 to 10 µM for 24 h in SW480 and C2C12 cultures) and selectively prevents CBP- but not p300-, dependent TCF/β-catenin transcriptional regulations both in cultures in vitro (5 to 25 µM in SW480, HCT116, rat RLT-6TN, and primary human IPF lung fibroblasts; 4 to 24 h) and in mice in vivo (5 mg/kg/day via osmotic pump infusion), while not affecting CBP-dependent transcriptional activities mediated by the AP-1 or CRE complex. Selective blockage of CBP/TCF/β-catenin complex formation by ICG-001 is shown to reactivate p300//TCF/β-catenin-mediated neuronal differentiation upon NGF stimulation in PS-1 L286V mutant-expressing PC-12 cultures (cell population with neurite outgrowth = 90% and 17% of wt PS-1 control level, respectively, with or without 10 µM ICG-001) and be efficacious in alleviating bleomycin- (0,08 units/50 µL/mouse; administered intranasally; Cat. No. 203401) induced pulmonary fibrosis in mice (5 mg/kg/day via osmotic pump infusion; 10 d treatment starting 1 d before bleomycin induction or 21 d treatment starting d 21 post bleomycin induction).
A cell-permeable pyrazinopyrimidinecarboxamide that competes against β-catenin for CBP (CREB-binding protein) binding and selectively prevents CBP- but not p300-, dependent TCF/β-catenin transcriptional regulations both in cultures in vitro (5 to 25 µM) and in mice in vivo (5 mg/kg/day via osmotic pump infusion), while not affecting CBP-dependent transcriptional activities mediated by the AP-1 or CRE complex. Shown to reactivate p300//TCF/β-catenin-mediated neuronal differentiation upon NGF stimulation in PS-1 L286V mutant-expressing PC-12 cultures and be efficacious in alleviating bleomycin- (0,08 units/50 µL/mouse; administered intranasally; Cat. No. 203401) induced pulmonary fibrosis in mice.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CREB-binding protein
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Yan, D., et al. 2012. J. Biol. Chem.287, 8598.
Henderson, W.R. Jr, et al. 2010. Proc. Natl. Acad. Sci. USA.107, 14309.
Eguchi, M., et al. 2005. Med Chem.1, 467.
Ma, H., et al. 2005. Oncogene.24, 3619.
Teo, J.L., et al. 2005. Proc. Natl. Acad. Sci. USA.102, 12171.
Emami, K.H., et al. 2004. Proc. Natl. Acad. Sci. USA.101, 12682.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service