Skip to Content
Merck
All Photos(1)

Key Documents

475915

Sigma-Aldrich

Anti-Myeloperoxidase Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPO(4353)

General description

Rabbit polyclonal antibody purified by ammonium sulfate precipitation followed by DEAE-Sephacel ion exchange chromatography. Recognizes the myeloperoxidase protein.
Recognizes myeloperoxidase in human neutrophil granules.
This Anti-Myeloperoxidase Rabbit pAb is validated for use in ELISA, Immunoelectrophoresis for the detection of Myeloperoxidase.

Immunogen

Human
purified, human neutrophil myeloperoxidase

Application

ELISA (≥ 1:4000)

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Generates a single arc by immunoelectrophoresis against human neutrophil granule extract and purified MPO. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Maritati et al.
Frontiers in microbiology, 15, 1417049-1417049 (2024-06-24)
The accurate distinction between periprosthetic joint infections (PJI) and aseptic failures (AF) is of paramount importance due to differences in treatment. However, this could be challenging by using the current criteria. Various synovial fluid biomarkers are being assessed to improve
Tobias Deuse et al.
Nature, 509(7502), 641-644 (2014-04-22)
Despite the introduction of antiproliferative drug-eluting stents, coronary heart disease remains the leading cause of death in the United States. In-stent restenosis and bypass graft failure are characterized by excessive smooth muscle cell (SMC) proliferation and concomitant myointima formation with
Sean E Gill et al.
American journal of respiratory cell and molecular biology, 55(2), 243-251 (2016-03-05)
Although neutrophils play critical roles in innate immunity, in excess these cells cause severe tissue damage. Thus, neutrophil activation must be tightly regulated to prevent indiscriminant damage. Previously, we reported that mice lacking matrix metalloproteinase (MMP) 7 are protected from
Volker Rudolph et al.
Nature medicine, 16(4), 470-474 (2010-03-23)
Observational clinical and ex vivo studies have established a strong association between atrial fibrillation and inflammation. However, whether inflammation is the cause or the consequence of atrial fibrillation and which specific inflammatory mediators may increase the atria's susceptibility to fibrillation
Kashish Manchanda et al.
Arteriosclerosis, thrombosis, and vascular biology, 38(8), 1859-1867 (2018-06-16)
Objective- The leukocyte heme-enzyme MPO (myeloperoxidase) exerts proinflammatory effects on the vascular system primarily linked to its catalytic properties. Recent studies have shown that MPO, depending on its cationic charge, mediates neutrophil recruitment and activation. Here, we further investigated MPO's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service