Skip to Content
Merck
All Photos(1)

Key Documents

474780

Sigma-Aldrich

MG-115

Potent reversible proteasome inhibitor (Ki = 21 nM and 35 nM for 20S and 26S proteasome, respectively).

Synonym(s):

MG-115, Carbobenzoxy-L-leucyl-L-leucyl-L-norvalinal, Z-LL-Nva-CHO, Proteasome Inhibitor XII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H39N3O5
CAS Number:
Molecular Weight:
461.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N([C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC)C=O)C(=O)OCc1ccccc1

InChI

1S/C25H39N3O5/c1-6-10-20(15-29)26-23(30)21(13-17(2)3)27-24(31)22(14-18(4)5)28-25(32)33-16-19-11-8-7-9-12-19/h7-9,11-12,15,17-18,20-22H,6,10,13-14,16H2,1-5H3,(H,26,30)(H,27,31)(H,28,32)/t20-,21-,22-/m0/s1

InChI key

QEJRGURBLQWEOU-FKBYEOEOSA-N

General description

Potent reversible proteasome inhibitor (Ki = 21 nM and 35 nM for 20S and 26S proteasome, respectively). Specifically inhibits the chymotrypsin-like activity of the proteasome and induces apoptosis in Rat-1 and PC12 cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
20S proteasome
Product does not compete with ATP.
Reversible: yes
Target Ki: 21 nM and 35 nM for 20S and 26S proteasome, respectively

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Leu-Leu-Nva-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lopes, U.G., et al. 1997. J. Biol. Chem. 272, 12893.
Lee, D.H., and Goldberg, A.L. 1996. J. Biol. Chem.271, 27280.
Palombella, V.J., et al. 1994. Cell 78, 773. Rock, K.L., et al. 1994. Cell 78, 761.
Vinitsky, A., et al. 1992. Biochemistry 31, 9421.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service