Skip to Content
Merck
All Photos(1)

Key Documents

407135-M

Millipore

Caged Ins(1,4,5)P3, Trisodium Salt

Photolabile derivative of Ins(1,4,5)P₃ that is not biologically active and is not metabolized by endogenous phosphatase.

Synonym(s):

Caged Ins(1,4,5)P3, Trisodium Salt, D- myo-Inositol 1,4,5-Trisphosphate, P 4(5)-1-(2-Nitrophenyl)ethyl Ester, 3Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO17P3 · 3Na
Molecular Weight:
635.19
UNSPSC Code:
12352211
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

off-white

solubility

water: 1 mg/mL

storage temp.

−20°C

General description

Extinction coefficient (265 nm, H2O): 4240 M-1cm-1.
Note: 1 set = 10 x 10 µg.
Photolabile derivative of Ins(1,4,5)P3 that is not biologically active and is not metabolized by endogenous phosphatase (extinction coefficient: 4240 M-1cm-1; 265 nm, H2O). A mixture of two isomers with a cage on either the 4 or 5 position. Can be selectively released by irradiation with a short pulse of light of about 360 nm to produce a concentration jump of Ins(1,4,5)P3.

Biochem/physiol Actions

Primary Target
Photolabile derivative of Ins(1,4,5)P3 that is not biologically active

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Walker, J.W., et al. 1987. Nature327, 249.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service