Skip to Content
Merck
All Photos(1)

Documents

401211

Sigma-Aldrich

Goat Anti-Mouse IgG, H & L Chain Specific

Calbiochem®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoaffinity purified goat polyclonal antibody. Recognizes mouse IgG heavy and light chains.
This Goat Anti-Mouse IgG, H & L Chain Specific is validated for use in Immunoelectrophoresis, Immunoprecipitation for the detection of Mouse IgG, H & L Chain Specific.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Monospecific for mouse immunoglobulin G heavy and light chains as determined by immunoelectrophoresis against normal mouse serum. Comparison of immunoprecipitation analysis with total protein determines that product contains 95-100% active antibody. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuo Liu et al.
Oncology reports, 48(3) (2022-07-21)
The increasing morbidity and high mortality of intrahepatic cholangiocarcinoma (ICC) has led to the urgent need for new diagnostics and therapeutics. Liver kinase B1 (LKB1) exerts a tumor suppressor role in multiple malignances, while its regulatory role in exosomes secreted
Chengshan Xu et al.
Oncotarget, 7(47), 77764-77776 (2016-10-27)
Histone H2AX is a tumor suppressor protein that plays an important role in apoptosis. However, the mechanism underlying the association of H2AX with apoptosis in cancer cells remains elusive. Here, we showed that H2AX knockdown in lung cancer A549 cells
Yong Gao et al.
BioMed research international, 2018, 3842753-3842753 (2018-06-05)
Peroxisome proliferator-activated receptor α (PPARα) is a member of the nuclear hormone receptor superfamily and functions as a transcription factor. Previous work showed that PPARα plays multiple roles in lipid metabolism in tissues such as cardiac and skeletal muscle, liver
Diana C Muñoz-Lasso et al.
Scientific reports, 10(1), 5207-5207 (2020-04-07)
Abnormalities in actin cytoskeleton have been linked to Friedreich's ataxia (FRDA), an inherited peripheral neuropathy characterised by an early loss of neurons in dorsal root ganglia (DRG) among other clinical symptoms. Despite all efforts to date, we still do not
Ronggang Li et al.
Molecular medicine reports, 20(6), 4893-4904 (2019-10-23)
Theaflavin has been proven to own strong antioxidative capacity; however, the molecular mechanism underlying its protective effect against cerebral ischemia‑reperfusion (I/R) injury remains unclear. Therefore, the present study was designed to elucidate the neuroprotective effects of theaflavin on cerebral I/R

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service