Skip to Content
Merck
All Photos(1)

Key Documents

400046

Sigma-Aldrich

Hydroxyurea

≥98% (HPLC), solid, RNR inhibitor, Calbiochem

Synonym(s):

Hydroxyurea, Ribonucleotide Reductase Inhibitor I, RNR Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4N2O2
CAS Number:
Molecular Weight:
76.05
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Hydroxyurea, Anti-neoplastic agent that blocks DNA replication by inhibiting ribonucleotide reductase.

Quality Level

description

Merck USA index - 14, 4848

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(O)C(=O)N

InChI

1S/CH4N2O2/c2-1(4)3-5/h5H,(H3,2,3,4)

InChI key

VSNHCAURESNICA-UHFFFAOYSA-N

General description

Anti-neoplastic agent that blocks DNA replication by inhibiting ribonucleotide reductase. Induces apoptosis in chronic myelogenous leukemia lymphocytes. Reported to enhance the radiation effects in pancreatic adenocarcinoma cells. Releases NO in the presence of oxidizing agents.
Antineoplastic agent. Blocks DNA replication by inhibiting ribonucleotide reductase. Induces apoptosis in chronic myelogenous leukemia lymphocytes.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inhibiting ribonucleotide reductase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 2 months at 4°C.

Other Notes

Leyden, D., et al. 2000. Anticancer Res.20, 133.
Tsuchiya, K., et al. 1999. Free Radic. Biol. Med. 27, 347.
Anand, S., et al. 1995. Cancer Lett.88, 101.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Melissa Seman et al.
Cell reports, 42(11), 113428-113428 (2023-11-12)
H3K9 methylation (H3K9me) marks transcriptionally silent genomic regions called heterochromatin. HP1 proteins are required to establish and maintain heterochromatin. HP1 proteins bind to H3K9me, recruit factors that promote heterochromatin formation, and oligomerize to form phase-separated condensates. We do not understand
Xiaoju Hu et al.
NAR cancer, 4(3), zcac023-zcac023 (2022-07-29)
The histone methyltransferase KMT2C is among the most frequently mutated epigenetic modifier genes in cancer and plays an essential role in MRE11-dependent DNA replication fork restart. However, the effects of KMT2C deficiency on genomic instability during tumorigenesis are unclear. Analyzing
Kaisa Ikkala et al.
Scientific reports, 12(1), 161-161 (2022-01-09)
The cornea, transparent and outermost structure of camera-type eyes, is prone to environmental challenges, but has remarkable wound healing capabilities which enables to preserve vision. The manner in which cell plasticity impacts wound healing remains to be determined. In this
P Logan Schuck et al.
The Journal of biological chemistry, 297(3), 101026-101026 (2021-08-03)
Sister chromatid cohesion (SCC), the pairing of sister chromatids after DNA replication until mitosis, is established by loading of the cohesin complex on newly replicated chromatids. Cohesin must then be maintained until mitosis to prevent segregation defects and aneuploidy. However
Christopher Noyes et al.
Communications biology, 6(1), 65-65 (2023-01-19)
Human cancers often re-express germline factors, yet their mechanistic role in oncogenesis and cancer progression remains unknown. Here we demonstrate that DEAD-box helicase 4 (DDX4), a germline factor and RNA helicase conserved in all multicellular organisms, contributes to increased cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service