Skip to Content
Merck
All Photos(1)

Key Documents

370660

Sigma-Aldrich

Guanosine 3′,5′-cyclic Monophosphate, N²,2′-O-Dibutyryl-, Sodium Salt

Cell-permeable cGMP analog that preferentially activates protein kinase G.

Synonym(s):

Guanosine 3′,5′-cyclic Monophosphate, N²,2′-O-Dibutyryl-, Sodium Salt, Dibutyryl-cGMP, Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23N5O9P · Na
CAS Number:
Molecular Weight:
507.37
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

ethanol: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[P]1(=O)(O[C@@H]2[C@H](O[C@H]([C@@H]2OC(=O)CCC)[n]3c4nc(nc(c4nc3)O)NC(=O)CCC)CO1)[O-]

InChI

1S/C18H24N5O9P.Na/c1-3-5-10(24)20-18-21-15-12(16(26)22-18)19-8-23(15)17-14(31-11(25)6-4-2)13-9(30-17)7-29-33(27,28)32-13;/h8-9,13-14,17H,3-7H2,1-2H3,(H,27,28)(H2,20,21,22,24,26);/q;+1/p-1/t9-,13-,14-,17-;/m1./s1

InChI key

MGBPJXVWDGGLKI-GBIKJYCISA-M

Related Categories

General description

Cell-permeable cGMP analog that preferentially activates protein kinase G. Inhibits thrombin stimulated arachidonic acid release in human platelets.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cell-permeable cGMP analog that preferentially activates protein kinase G.
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Sane, D.C., et al. 1989. Biochem. Biophys. Res. Commun.165, 708.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service