Skip to Content
Merck
All Photos(1)

Key Documents

345788

Sigma-Aldrich

β-Galactosidase, E. coli

Synonym(s):

β-Galactosidase, E. coli, β-D-Galactoside galactohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Escherichia coli

Quality Level

description

RTECS - LW5840000

form

lyophilized solid

specific activity

≥600 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 2%

shipped in

ambient

storage temp.

−20°C

General description

Native E. coli β-galactosidase. Inducible enzyme not found in eukaryotic cells that is used as a reporter gene. Used to assess efficiency of transfection. Also used in ELISA and immunocytochemical techniques. Histochemical substrates include X-Gal (Cat. No. 203782).
Native E. coli β-galactosidase. Inducible enzyme not found in eukaryotic cells that is used as a reporter gene. Used to assess efficiency of transfection. Also used in ELISA and immunocytochemical techniques. Histochemical substrates include X-Gal (Cat. No. 203782).

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol of p-nitrophenyl-β-galactoside per min at 37°C.

Other Notes

Zeleny, R., et al. 1997. Anal. Biochem.246, 96.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service