Skip to Content
Merck
All Photos(1)

Documents

239765

Sigma-Aldrich

Cycloheximide

InSolution, 100 mg/mL in DMSO, Suitable for cell culture

Synonym(s):

InSolution Cycloheximide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.77

Quality Level

form

solution

manufacturer/tradename

Calbiochem®

concentration

100 mg/mL in DMSO

solubility

DMSO: soluble

storage temp.

2-8°C

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

General description

A convenient ready-to-use form of Cycloheximide (Cat. No. 239763). An anti-fungal antibiotic that inhibits protein synthesis in eukaryotes but not in prokaryotes. Interacts directly with the translocase enzyme, interfering with the translocation step. Also inhibits cell-free protein synthesis in eukaryotes. Competitively inhibits hFKBP12 (Ki = 3.4 µM). Triggers apoptosis in HL-60 cells, T-cell hybridomas, Burkitt′s lymphoma cells, and a variety of other cell types, including rodent macrophages. Induces DNA fragmentation in macrophages, but inhibits DNA cleavage in rat thymocytes treated with thapsigargin, methylprednisolone, and ionomycin. Rapidly destroyed in alkaline solutions.

Biochem/physiol Actions

Primary Target
hFKBP12

Packaging

Packaged under inert gas

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Physical form

A 100 mg/ml solution of Cycloheximide (Cat. No. 239763) in DMSO (sterile-filtered).

Reconstitution

Following initial use, aliquot and refrigerate (4°C).

Other Notes

Christner, C., et al. 1999. J. Med. Chem.42, 3615.
Lu, Q. and Mellgreen,R.L. 1996. Arch. Biochem. Biophys.334, 175.
Chow, S.C., et al. 1995. Exp. Cell Res.216, 149.
Waring, P. 1990. J. Biol. Chem.265, 14476.
Obrig, T.G., et al. 1971. J. Biol. Chem.246, 174.
Pestka, S. 1971. Annu. Rev. Microbiol.25, 487.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joanan Lopez-Morales et al.
ACS synthetic biology, 12(2), 419-431 (2023-02-03)
Yeast surface display is a valuable tool for protein engineering and directed evolution; however, significant variability in the copy number (i.e., avidity) of displayed variants on the yeast cell wall complicates screening and selection campaigns. Here, we report an engineered
Zachary J Waldrip et al.
The Journal of biological chemistry, 297(4), 101209-101209 (2021-09-26)
DNA-dependent protein kinase catalytic subunit (DNA-PKcs) is known primarily for its function in DNA double-stranded break repair and nonhomologous end joining (NHEJ). However, DNA-PKcs also has a critical yet undefined role in immunity impacting both myeloid and lymphoid cell lineages
Luyao Wu et al.
Journal of translational medicine, 22(1), 201-201 (2024-02-25)
Although the long-term prognosis of papillary thyroid cancer (PTC) is favorable, distant metastasis significantly compromises the prognosis and quality of life for patients with PTC. The Cadherin family plays a pivotal role in tumor metastasis; however, the involvement of Cadherin
Zhiqian Zhang et al.
Molecular cell, 83(18), 3377-3392 (2023-09-23)
The ubiquitin-proteasome system plays a critical role in biology by regulating protein degradation. Despite their importance, precise recognition specificity is known for a few of the 600 E3s. Here, we establish a two-pronged strategy for identifying and mapping critical residues
Morag Rose Hunter et al.
Small methods, 7(9), e2201695-e2201695 (2023-06-15)
Poor understanding of intracellular delivery and targeting hinders development of nucleic acid-based therapeutics transported by nanoparticles. Utilizing a siRNA-targeting and small molecule profiling approach with advanced imaging and machine learning biological insights is generated into the mechanism of lipid nanoparticle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service