Skip to Content
Merck
All Photos(1)

Documents

20-103

Sigma-Aldrich

Phosphate Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

shipped in

wet ice

Application

For use in phosphatase assay kits.
Research Category
All

Quality

Routinely evaluated by Malachite Green Assay.

Physical form

1mM potassium phosphate monobasic

Storage and Stability

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Historical and contemporary correlates of syphilis and cancer.
A M Michalek,M C Mahoney,C C McLaughlin,D Murphy,B B Metzger
International Journal of Epidemiology null
K Kruus et al.
Applied microbiology and biotechnology, 44(3-4), 399-404 (1995-12-01)
CelS is the most abundant subunit and an exoglucanase component of the Clostridium thermocellum cellulosome, multicomponent cellulase complex. The product inhibition pattern of CelS was examined using purified recombinant CelS (rCelS) produced in Escherichia coli. The rCelS activity on cellopentaose
Immunochemical mapping of domains in human interleukin 4 recognized by neutralizing monoclonal antibodies.
L Ramanathan,R Ingram,L Sullivan,R Greenberg,R Reim,P P Trotta,H V Le
Biochemistry null
Selective proteolytic cleavage of recombinant human interleukin 4. Evidence for a critical role of the C-terminus.
H V Le,G F Seelig,R Syto,L Ramanathan,W T Windsor,D Borkowski,P P Trotta
Biochemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service