Skip to Content
Merck
All Photos(1)

Key Documents

1.93216

Supelco

Copper(II) sulfate pentahydrate

for analysis EMPARTA® ACS

Synonym(s):

Copper(II) sulfate pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO4S · 5H2O
CAS Number:
Molecular Weight:
249.69
MDL number:
UNSPSC Code:
12352302
grade:
ACS reagent

grade

ACS reagent

storage temp.

room temp

SMILES string

[Cu+2].[S](=O)(=O)([O-])[O-].O.O.O.O.O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93216.0521: 500 g, Minimum order quantity: 1
1.93216.5021: 5 kg, Minimum order quantity: 1
1.93216.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service