Skip to Content
Merck
All Photos(2)

Documents

1.88020

Supelco

CombiSolvent Oil

solvent for volumetric Karl Fischer titration with one component reagents for oils, Aquastar®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601
NACRES:
NA.21

product name

CombiSolvent Oil, Solvent for volumetric Karl Fischer titration with one component reagents for oils Aquastar®

Quality Level

product line

Aquastar®

form

liquid

reaction suitability

reaction type: volumetric

technique(s)

Karl-Fischer titration: suitable
titration: suitable

pH

5.6 (20 °C in H2O)

kinematic viscosity

0.67 cSt(40 °C)

transition temp

flash point 6 °C

density

0.86 g/cm3 at 20 °C

format

single component solution

storage temp.

15-25°C

Analysis Note

Application test: passes test

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F

Flash Point(C)

6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service