1.31200
HY-RiSE® Color Hygiene Test Strip
Rapid colorimetric test for visual determination of surface cleanliness and hand hygiene
Synonym(s):
Colorimetric assay, Paper strip test
About This Item
Recommended Products
material
paper (Test Strips: individually sealed in aluminum foil)
plastic (box)
plastic bottle (Reagent Bottles with colored screw caps)
form
liquid
shelf life
24 mo. at 20‑25 °C
packaging
box of 50 strips (individually sealed)
pkg of Reagent A (wetting solution, white screw cap, 2.5 ml)
pkg of Reagent B (substrate solution, yellow screw cap, 2.5 ml)
pkg of Reagent C (enzyme solution, blue screw cap, 2.0 ml)
manufacturer/tradename
HY-RISE®
storage condition
protect from light
application(s)
clinical
cosmetics
food and beverages
industrial hygiene (monitoring)
microbiology
surface monitoring
compatibility
for use with HY-RISE®
detection method
colorimetric
storage temp.
2-8°C
Related Categories
General description
Test results are available in 4 - 5 minutes and require no equipment. Results are presented as a color change on the test strip, making it easy to determine clean (Pass) and dirty (Fail) results. Surface cleanliness is determined by measuring product residues, e.g. from food and beverages, left on surfaces after cleaning. Measurements made with this test strip can provide early warning of possible contamination on specific surface areas allowing immediate corrective action. On visibly clean surfaces the test can reveal the presence of product residues invisible to the eye, therefore indicating the hidden potential for microbial growth.
The HY-RiSE® color hygiene test measures the product residues in the form of nicotinamide adenine dinucleotides (NAD, NADH) and their phosphates (NADP, NADPH). Regular use of the HY-RiSE® provides a simple and effective means for monitoring standards of cleaning as part of an integrated hygiene monitoring program.
Application
Linkage
Analysis Note
Appearance
- reagent A:
clear, colourless
Reagent B
- reagent B: clear, yellowish
Colony count
- reagent B:
≤ 10 CFU/ml
Reagent C
Appearance
- reagent C:
clear, yellowish
Colony count
- reagent C:
≤ 10 CFU/ml
performance test
Performance test: conforms
Legal Information
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service