Skip to Content
Merck
All Photos(1)

Key Documents

1.20595

Millipore

CSB Selective Supplement (Vancomycin)

Synonym(s):

CSB Selective Supplement (Vancomycin)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · xHCl
CAS Number:
Molecular Weight:
1449.25 (free base basis)
MDL number:
UNSPSC Code:
41106212
EC Index Number:
215-772-6
NACRES:
NA.74

Agency

ISO

Quality Level

form

lyophilized powder

pH

2.5-4.5 (20 °C, 50 g/L in H2O)

application(s)

food and beverages
microbiology

shipped in

ambient

storage temp.

2-8°C

suitability

Cronobacter spp.

SMILES string

Clc1c2ccc(c1)[C@H]([C@@H]3NC(=O)[C@H](NC(=O)[C@@H]6NC(=O)[C@@H](NC(=O)[C@@H]([C@@H](c7cc(c(cc7)Oc8c(c(cc6c8)O2)O[C@@H]9O[C@@H]([C@H]([C@@H]([C@H]9O[C@@H]%10O[C@H]([C@H]([C@](C%10)(N)C)O)C)O)O)CO)Cl)O)NC(=O)[C@H](NC)CC(C)C)CC(=O)N)c4cc(c(cc4)O)c5c(cc(cc5O)

InChI

1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)

InChI key

LCTORFDMHNKUSG-XTTLPDOESA-N

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service