Skip to Content
Merck
All Photos(1)

Key Documents

118502

Sigma-Aldrich

ATM Kinase Inhibitor

InSolution, ≥95%

Synonym(s):

InSolution ATM Kinase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17NO3S2
Molecular Weight:
395.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable disubstituted pyranone compound that acts as a potent and ATP-competitive inhibitor of ATM kinase (IC50 = 13 nM; Ki = 2.2 nM). Displays excellent selectivity over other PIKK family kinases (IC50 = 2.5, 9.3, 16.6 µM for DNA-PK, mTOR, PI 3-K, respectively; IC50 >100 µM for PI 4-K and ATR) and exhibits little activity towards a panel of 60 other kinases even at concentrations as high as 10 µM. Inhibits ATM-dependent cellular protein phosphorylation following ionizing radiation (IR) and sensitizes cells with wild-type ATM, but not mutant ATM, to the cytotoxic effects of IR and DNA-damaging agents.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (2 mg/506 µl) solution of ATM Kinase Inhibitor (Cat. No. 118500) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Pereg, Y., et al. 2005. Proc. Natl. Acad. Sci. USA102, 5056.
Lau, A., et al. 2005. Nat. Cell Biol.7, 493.
Hickson, I., et al. 2004. Cancer Res.64, 9152.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaron Pereg et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(14), 5056-5061 (2005-03-25)
Maintenance of genomic stability depends on the DNA damage response, an extensive signaling network that is activated by DNA lesions such as double-strand breaks (DSBs). The primary activator of the mammalian DSB response is the nuclear protein kinase ataxia-telangiectasia, mutated
Alan Lau et al.
Nature cell biology, 7(5), 493-500 (2005-04-19)
Chemotherapy that is used to treat human immunodeficiency virus type-1 (HIV-1) infection focuses primarily on targeting virally encoded proteins. However, the combination of a short retroviral life cycle and high mutation rate leads to the selection of drug-resistant HIV-1 variants.
Ian Hickson et al.
Cancer research, 64(24), 9152-9159 (2004-12-18)
The serine/threonine protein kinase ATM signals to cell cycle and DNA repair components by phosphorylating downstream targets such as p53, CHK2, NBS1, and BRCA1. Mutation of ATM occurs in the human autosomal recessive disorder ataxia-telangiectasia, which is characterized by hypersensitivity
Consuelo Pitolli et al.
Journal of experimental & clinical cancer research : CR, 42(1), 214-214 (2023-08-21)
Medulloblastoma (MB) is the most common cerebellar malignancy during childhood. Among MB, MYC-amplified Group 3 tumors display the worst prognosis. MYC is an oncogenic transcription factor currently thought to be undruggable. Nevertheless, targeting MYC-dependent processes (i.e. transcription and RNA processing
Estelle Vincendeau et al.
Nature communications, 13(1), 3707-3707 (2022-06-29)
SHLD1 is part of the Shieldin (SHLD) complex, which acts downstream of 53BP1 to counteract DNA double-strand break (DSB) end resection and promote DNA repair via non-homologous end-joining (NHEJ). While 53BP1 is essential for immunoglobulin heavy chain class switch recombination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service