Skip to Content
Merck
All Photos(1)

Documents

1.18306

Supelco

Heptane-1-sulfonic acid sodium salt

for ion pair chromatography LiChropur

Synonym(s):

Heptane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NaO3S
CAS Number:
Molecular Weight:
202.25
MDL number:
UNSPSC Code:
12352107
EC Index Number:
245-210-5
NACRES:
NB.21

Quality Level

Assay

≥99% (acidimetric)

technique(s)

ion pair chromatography: suitable

transmittance

200 nm, ≥70%
220 nm, ≥90%
250 nm, ≥98%
(0.005 M; 1 cm in H2O)

pH

5.5-7.5 (20 °C, 100 g/L in H2O)

mp

>300 °C

bulk density

391 kg/m3

storage temp.

15-25°C

InChI

1S/C7H16O3S.Na/c1-2-3-4-5-6-7-11(8,9)10;/h2-7H2,1H3,(H,8,9,10);/q;+1/p-1

InChI key

REFMEZARFCPESH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Heptane-1-sulfonicacid sodium salt is commonly employed as an ion-pair reagent in reversed-phaseliquid chromatography (RPLC) to improve the separation and retention ofanalytes.

Application

Heptane-1-sulfonic acid sodium salt can be used as a mobile phase component in high-performance liquid chromatographic methods to determine water-soluble vitamins.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service