Skip to Content
Merck
All Photos(1)

Key Documents

1.14670

Supelco

Chlorine Test Kit, colorimetric

0.10-2.0 mg/L (Cl₂), MQuant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Chlorine Test in freshwater and seawater, colorimetric, 0.10-2.0 mg/L (Cl2), for use with MQUANT®

specific analyte(s)

chlorine

Quality Level

measuring range

0.10-2.0 mg/L (Cl2)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card 0.10 - 0.25 - 0.5 - 1.0 - 2.0 mg/l Cl₂ MQuant®
This MQuant® Chlorine test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card. The reagents are added to the sample, resulting in a color reaction. The concentration is determined by allocating the color to a value on a reference scale.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service