1.12356
Tellurium dioxide
99.5+
Synonym(s):
Tellurium dioxide, Tellurium(IV) oxide
About This Item
Quality Level
form
solid
potency
>2000 mg/kg LD50, oral (Rat)
mp
733 °C
density
6.02 g/cm3 at 20 °C
storage temp.
no temp limit
SMILES string
[Te](=O)=O
InChI
1S/O2Te/c1-3-2
InChI key
LAJZODKXOMJMPK-UHFFFAOYSA-N
Related Categories
Application
- Facile sonochemical synthesis of tellurium and tellurium dioxide nanoparticles: This study presents a method for synthesizing tellurium nanostructures by reducing TeCl4 to Te using ultrasonic irradiation in methanol, also exploring the oxidation process to produce tellurium dioxide (Arab, Mousavi-Kamazani, 2017).
- Ultrathin tellurium dioxide: emerging direct bandgap semiconductor: This article discusses the properties of monolayer tellurium dioxide as a new 2D semiconducting oxide, highlighting its direct bandgap and high mobility, which are significant for various applications (Guo et al., 2018).
Analysis Note
Identity (ICP-OES): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Lact. - Repr. 1B - Skin Sens. 1B
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service