Skip to Content
Merck
All Photos(2)

Key Documents

1.11732

Sigma-Aldrich

Trypan blue (C.I. 23850)

for microscopy

Synonym(s):

Trypan Blue, Benzamine blue, Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium salt, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Direct blue 14

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H24N6O14S4Na4
CAS Number:
Molecular Weight:
960.81
Colour Index Number:
23850
Beilstein:
4360496
MDL number:
UNSPSC Code:
41116130
EC Index Number:
200-786-7

Quality Level

form

solid

potency

6200 mg/kg LD50, oral (Rat)

composition

Dye, ≥80% spectrophotometric assay

pH

9.8 (20 °C, 10 g/L in H2O)

mp

>300 °C (lit.)

solubility

10 g/L

bulk density

800 kg/m3

εmax

650-820 at 590-600 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(ccc1N=Nc2c(O)c3c(N)cc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O)-c4ccc(N=Nc5c(O)c6c(N)cc(cc6cc5S([O-])(=O)=O)S([O-])(=O)=O)c(C)c4

InChI

1S/C34H28N6O14S4.4Na/c1-15-7-17(3-5-25(15)37-39-31-27(57(49,50)51)11-19-9-21(55(43,44)45)13-23(35)29(19)33(31)41)18-4-6-26(16(2)8-18)38-40-32-28(58(52,53)54)12-20-10-22(56(46,47)48)14-24(36)30(20)34(32)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4

InChI key

GLNADSQYFUSGOU-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Trypan blue (C.I. 23850) for microscopy, is a dry dye for the preparation of a color staining solution, staining histological sample material of human origin, e.g. trichrome staining in collagenous connective tissue.
The dye is a CE certified IVD product and has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining.

Analysis Note

Dye content (spectrophotometrically): ≥ 80 %
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 590 - 600 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water): 650 - 820
TLC-Test: passes test
Loss on drying (110 °C): ≤ 15 %
Suitability for microscopy: passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service