Skip to Content
Merck
All Photos(2)

Key Documents

1.10084

Supelco

Peracetic Acid Test

greener alternative

colorimetric, 5-50 mg/L (peracetic acid), for use with MQuant® StripScan App, MQuant®

Synonym(s):

Peracetic Acid chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

peracetic acid

reaction suitability

reaction type: Redox Reactions

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

5-50 mg/L (peracetic acid)

compatibility

for use with MQuant® StripScan App

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 5 - 10 - 20 - 30 - 50 mg/L (peracetic acid), MQuant®

Peracetic acid is a frequently used disinfectant. The MQuant® Peracetic acid test strips are applicable – even in the presence of hydrogen peroxide – to test for the absence of peracetic acid residues after rinsing cycles. Peracetic acid reacts with an aromatic amine to form a blue dye. The concentration of peracetic acid is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the peracetic acid concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® test.

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.This product has benefited from:
  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Linkage

Replaces: 10084-1; 10084

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service