Skip to Content
Merck
All Photos(2)

Key Documents

1.10024

Supelco

Ammonium Test Strips, colorimetric

greener alternative

10-400 mg/L (NH₄⁺), MQuant®, MQuant® StripScan App

Synonym(s):

Ammonium chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Ammonium Test, colorimetric, 10-400 mg/L (NH4+), for use with MQuant® StripScan App, MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

ammonium

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

10-400 mg/L (NH4+)

compatibility

for use with MQuant® StripScan App

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric with test strips and reagent 10 - 30 - 60 - 100 - 200 - 400 mg/l NH₄⁺ MQuant®

The test is suitable for determination potentially harmful pollution in surface and ground water, which are caused by ammonium compounds. Depending on the pH level ammonia is formed, which is a potent toxin for fish. In agriculture the MQuant® test strips can be used to measure the ammonium nitrogen content of various types of manure and compost after sample preparation. Using these test strips, the ammonium concentration is measured semiquantitatively by visual comparison of the reaction zone on the test strip with the fields of the color scale. All necessary reagents for the test are already included for a fast and easy analysis. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the ammonium concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Linkage

Replaces: 10024-1; 10024

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service