Skip to Content
Merck
All Photos(1)

Key Documents

1.08774

Supelco

Zinc

dust particle size < 63 µm EMPLURA®

Synonym(s):

Zinc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
EC Index Number:
231-175-3
NACRES:
NA.21
Assay:
≥95% metallic Zn basis (manganometric)
≥97.0% Zn basis (complexometric)
form:
powder
Pricing and availability is not currently available.

vapor pressure

1.33 hPa ( 487 °C)

Quality Level

product line

EMPLURA®

Assay

≥95% metallic Zn basis (manganometric)
≥97.0% Zn basis (complexometric)

form

powder

autoignition temp.

460 °C

impurities

≤0.5% Substances insoluble in nitric acid

bp

908 °C/1013 hPa

mp

420 °C

density

7.14 g/cm3 at 20 °C

bulk density

1800‑2700 kg/m3

cation traces

Cd: ≤0.2%
Cu: ≤0.005%
Fe: ≤0.01%
Pb: ≤1.0%
Sn: ≤0.03%

storage temp.

2-30°C

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Analysis Note

Assay (complexometric, total Zn): ≥ 97.0 %
Assay (manganometric, metall. Zn): ≥ 95.0 %
Substances insoluble in nitric acid: ≤ 0.5 %
Cd (Cadmium): ≤ 0.2 %
Cu (Copper): ≤ 0.005 %
Fe (Iron): ≤ 0.01 %
Pb (Lead): ≤ 1.0 %
Sn (Tin): ≤ 0.03 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service