Skip to Content
Merck
All Photos(2)

Key Documents

1.06639

Supelco

Sodium sulfate

anhydrous granulated for organic trace analysis EMSURE®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-820-9
NACRES:
NA.21
Assay:
≥99.0% (acidimetric)
form:
solid
solubility:
200 g/L

Quality Level

product line

EMSURE®

Assay

≥99.0% (acidimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

loss

≤ 0.5% loss on drying, 130 °C

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

solubility

200 g/L

density

2.68 g/mL at 25 °C (lit.)

bulk density

1400‑1600 kg/m3

cation traces

Fe: ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Appearance: small crystals
Assay (acidimetric): ≥ 99.0 %
Fe (Iron): ≤ 0.001 %
Loss on drying (130 °C): ≤ 0.5 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service