Skip to Content
Merck
All Photos(1)

Key Documents

1.05712

Millipore

MR-VP broth

Methyl-red VOGES-PROSKAUER broth, suitable for microbiology

Synonym(s):

Methyl-red VOGES-PROSKAUER broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

FDA-BAM

Quality Level

form

powder

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | metabolite detection: suitable
microbiological culture: suitable

pH

6.8-7.0 (37 °C, 17 g/L in H2O, after autoclaving)

solubility

17 g/L

bulk density

760 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Citrobacter spp.
Enterobacter spp.
Escherichia coli
Klebsiella spp.
Proteus spp.
Salmonella spp.
Yersinia spp.
coliforms

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 6.7 - 7.1
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): good to very good
Growth (Klebsiella pneumoniae ATCC 10031): good to very good
Growth (Enterobacter cloacae ATCC 13047 (WDCM 00083)): good to very good
Growth (Serratia marcescens ATCC 14756): good to very good
Methyl red (Escherichia coli ATCC 25922 (WDCM 00013)): +
Methyl red (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): +
Methyl red (Klebsiella pneumoniae ATCC 10031): +
Methyl red (Enterobacter cloacae ATCC 13047 (WDCM 00083)): -
Methyl red (Serratia marcescens ATCC 14756): +/-
VOGES-PROSKAUER (Escherichia coli ATCC 25922 (WDCM 00013)): -
VOGES-PROSKAUER (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): -
VOGES-PROSKAUER (Klebsiella pneumoniae ATCC 10031): -
VOGES-PROSKAUER (Enterobacter cloacae ATCC 13047 (WDCM 00083)): +
VOGES-PROSKAUER (Serratia marcescens ATCC 14756): +
Incubation: 48 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service