Skip to Content
Merck
All Photos(1)

Key Documents

1.05512

Supelco

Iron(III) chloride solution

10% Fe, for analysis EMSURE®

Synonym(s):

Ferric Chloride Aqueous Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.21
form:
liquid
Pricing and availability is not currently available.

Product Name

Iron(III) chloride solution (10% Fe), for analysis EMSURE®

Quality Level

product line

EMSURE®

form

liquid

concentration

10.0-10.3% (complexometric)

impurities

≤0.5% Free acid (as HCl)

pH

<1 (20 °C in H2O)

density

1.29 g/cm3 at 20 °C

anion traces

phosphate (PO43-): ≤0.005%

cation traces

As: ≤0.0005%
Fe2+: ≤0.005%
N: ≤0.005%
heavy metals (as Pb): ≤0.005%

storage temp.

15-25°C

Analysis Note

Assay (complexometric): 10.0 - 10.3 %
Free acid (as HCl): ≤ 0.5 %
As (Arsenic): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.005 %
Heavy metals as Pb (as lead): ≤ 0.005 %
Fe II (Iron II): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Does 10% Fe mean 10 g Fe/ mL?

    1 answer
    1. A percent solution refers to grams per 100 milliliter. The 10% indicates the product contains 10 grams Iron per 100ml volume.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service