Skip to Content
Merck

104662

SAFC

Cellvento® ModiFeed Sial+ COMP

greener alternative

Chemically defined cell culture feed - EMPROVE® CCM

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

cell culture | mammalian: suitable

greener alternative category

storage temp.

2-8°C

General description

Cellvento® ModiFeed Sial+ COMP is part of the ModiFeed platform. It is a highly concentrated, one-part, pH neutral feed added to replenish depleted nutrients required for cellular function and to extend the production phase in fed-batch mode, while simultanously maximizing sialylation on the protein of interest. It is recommended combination with the production media Cellvento® 4CHO and EX-CELL® Advanced CHO or other production media.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Application

Control of protein glycosylation, such as sialylation, while maximizing productivity is key for successful therapeutic protein development and production. N-glycoslyations of IgG Fc, particularly sialylation, has been reported to exert significant anti-inflammatory effects. High sialylation levels can help to increase stability, safety, and efficacy of the protein of interest. Cellvento® ModiFeed Sial+ COMP was developed to increase sialylation while maintaining or improving titer in CHO fed-batch processes

Features and Benefits

  • Reduced complexity in fed-batch process
  • Complete feed maximizing galactosylation
  • Highly concentrated to reduce required feed volume
  • Compacted format for conventient handling
  • Higher liquid feed stability at room temperature

Quantity

Formulated to contain 155.8 grams of powder per liter of feed.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service